Learn More
Electron Microscopy Sciences Hinge Microscope Slide Box with 100 slide capacity, Red
SDP
Supplier:  Electron Microscopy Sciences 71470R
Encompass_Preferred
Made from durable polystyrene, the slide box is unaffected by humidity and is thoroughly insect proof. Each box is secured with a rust-resistant nickel-plated clasp and hinge pin. The cork lining prevents damage to the stored slides. To ensure proper slide identification, each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box. To save space the boxes are stackable. Each box stores up to 100 3x1" or 25x75 mm microscope slides. Each box measures 8.75" x 6.75" x 3.375" (222x172x35 mm).
Color: Red
Catalog No. 50-189-9607