Learn More
Beckman Coulter Ultra Floor Rotor Accessories
Catalog No. NC1555175
Encompass
The Beckman Coulter Ultra-Floor Rotor Accessory is a floor stand designed to hold Beckman Coulter ultracentrifuge rotors when not in use. It is constructed of coated steel with rubber bumpers to protect the rotor. The stand has a wide base for stability and rubber feet to prevent sliding. It can accommodate rotors up to 30 inches in diameter and manage rotors weighing up to 200 pounds. The stand allows easy access to rotors and simplifies loading and unloading rotors from the ultracentrifuge.
Catalog No. NC1555175 Supplier Beckman Coulter Supplier No. 330070
The Beckman Coulter Ultra-Floor Rotor Accessory is a floor stand designed to hold Beckman Coulter ultracentrifuge rotors when not in use. It is constructed of coated steel with rubber bumpers to protect the rotor. The stand has a wide base for stability and rubber feet to prevent sliding. It can accommodate rotors up to 30 inches in diameter and manage rotors weighing up to 200 pounds. The stand allows easy access to rotors and simplifies loading and unloading rotors from the ultracentrifuge.