Learn More
Andwin Scientific HINGED 100-SLIDE BOX 10/PK
SDP
Supplier:  Andwin Scientific 71471W
Encompass_Preferred
With a cork lining, the Secured Hinge Microscope Slide Boxes are able to securely hold up to 100 3" x 1" or 25 x 75mm microscope slides. This is constructed from sturdy polystyrene (with heavy walls), can handle humidity, and thoroughly insect proof. Each slot is labeled with a number that resembles the slide inventory sheet provided on the inside cover of the box. The boxes are stackable, which allows users to save bench space. This product is sold in a pack of 10 white boxes.
Catalog No. 50-931-113