Learn More
Hach Company Hinged clamp 304 SS-2in
Catalog No. 5000207273 Shop All Hach Company Products
Encompass
2-inch Sanitary Clamp for 3455-series and PC/RC series sensors Stainless Steel 316 L Material
Catalog No. 50-002-07273 Supplier Hach Company Supplier No. 9H1132
2-inch Sanitary Clamp for 3455-series and PC/RC series sensors Stainless Steel 316 L Material