Learn More
MTC Bio FlipTop™ Carboard freezer box w/ attached hinged lid, 3 inch, 100-place, white, 50/cs
Catalog No. 502268487
Encompass_Preferred
MTC Bio’s FlipTop™ hinged lid boxes are made of recyclable cardboard, with a liquid resistant outer coating. They are shipped fully assembled with 81 place or 100 place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, etc). Available in white and 5 individual colors, plus a “Rainbow Pack” of 5 assorted colors. Useful temperature range: -196° to +121°C. 5 1/4 x 5 1/4 x 3 inches.
This item is not returnable. View return policy
Catalog No. 50-226-8487 Supplier MTC Bio Supplier No. R3700
MTC Bio’s FlipTop™ hinged lid boxes are made of recyclable cardboard, with a liquid resistant outer coating. They are shipped fully assembled with 81 place or 100 place cardboard dividers. Each box has drain slots on the bottom surface and numbered grid markings in almost every conceivable location (top, bottom, sides, etc). Available in white and 5 individual colors, plus a “Rainbow Pack” of 5 assorted colors. Useful temperature range: -196° to +121°C. 5 1/4 x 5 1/4 x 3 inches.