Learn More
Research Products International Corp Storage Box, Hinged Lid, 100 Tube Capacity, Black, 5 per Case
SDP
Catalog No. 50476638
Encompass
Durable polypropylene box is designed to protect light sensitive samples. Plastic boxes will not deteriorate or saturate like cardboard boxes. Plastic box withstands repeated low temperature storage. Hinged lid provides convenient access to sample tubes. Accommodates all hinged and screw cap 1.5 - 2.0 ml micro-tubes.
Catalog No. 50-476-638 Supplier Research Products International Corp Supplier No. 145638
Durable polypropylene box is designed to protect light sensitive samples. Plastic boxes will not deteriorate or saturate like cardboard boxes. Plastic box withstands repeated low temperature storage. Hinged lid provides convenient access to sample tubes. Accommodates all hinged and screw cap 1.5 - 2.0 ml micro-tubes.