Learn More
Electron Microscopy Sciences Polyethylene Hinged Lid Containers, 1 oz, 1-1/2" height, 1½" diameter
SDP
Catalog No. 5030420
Encompass_Preferred
These hinged lid containers are ideal for use just about anywhere. They feature a secure snap closure with hinged lid that eliminates lost caps. Containers offer air and moisture resistance to contents. Manufactured from flexible FD approvable polyethylene resin.
Volume: 1 oz
Height: 1-1/2"
Diameter: 1½"
Catalog No. 50-304-20 Supplier Electron Microscopy Sciences Supplier No. 6423810
These hinged lid containers are ideal for use just about anywhere. They feature a secure snap closure with hinged lid that eliminates lost caps. Containers offer air and moisture resistance to contents. Manufactured from flexible FD approvable polyethylene resin.
Volume: 1 oz
Height: 1-1/2"
Diameter: 1½"