Learn More
Electron Microscopy Sciences Polyethylene Hinged Lid Containers, 0.25 oz, 1/2" height, 1½" diameter
SDP
Catalog No. 5030417
Encompass_Preferred
These hinged lid containers are ideal for use just about anywhere. They feature a secure snap closure with hinged lid that eliminates lost caps. Containers offer air and moisture resistance to contents. Manufactured from flexible FD approvable polyethylene resin.
Volume: 0.25 oz
Height: 1/2"
Diameter: 1½"
Catalog No. 50-304-17 Supplier Electron Microscopy Sciences Supplier No. 6423801
These hinged lid containers are ideal for use just about anywhere. They feature a secure snap closure with hinged lid that eliminates lost caps. Containers offer air and moisture resistance to contents. Manufactured from flexible FD approvable polyethylene resin.
Volume: 0.25 oz
Height: 1/2"
Diameter: 1½"