Learn More
Electron Microscopy Sciences Polyethylene Hinged Lid Containers, 4 oz, 3-1/4" height, 2" diameter
SDP
Catalog No. 5030419
Encompass_Preferred
These hinged lid containers are ideal for use just about anywhere. They feature a secure snap closure with hinged lid that eliminates lost caps. Containers offer air and moisture resistance to contents. Manufactured from flexible FD approvable polyethylene resin.
Volume: 4 oz
Height: 3-1/4"
Diameter: 2"
Catalog No. 50-304-19 Supplier Electron Microscopy Sciences Supplier No. 6423820
These hinged lid containers are ideal for use just about anywhere. They feature a secure snap closure with hinged lid that eliminates lost caps. Containers offer air and moisture resistance to contents. Manufactured from flexible FD approvable polyethylene resin.
Volume: 4 oz
Height: 3-1/4"
Diameter: 2"