Learn More
Research Products International Corp Hinged Lid Box, Black, 50 Tube Capacity, 5 per Case
SDP
Catalog No. 50550190
Encompass
Durable polypropylene box is designed to protect light sensitive samples. Plastic box withstands repeated low temperature storage. Hinged lid provides convenient access to sample tubes. Accommondates all hinged and screw cap 1.5 - 2.0ml micro-tubes.
Catalog No. 50-550-190 Supplier Research Products International Corp Supplier No. 145637
Durable polypropylene box is designed to protect light sensitive samples. Plastic box withstands repeated low temperature storage. Hinged lid provides convenient access to sample tubes. Accommondates all hinged and screw cap 1.5 - 2.0ml micro-tubes.