Learn More
Elma™ Hinged Lids for Elmasonic Xtra ST Ultrasonic Cleaner
SDP
Catalog No. 08000121
Change view
Click to view available options
For Use With (Equipment):
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H
Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H
Ultrasonic Cleaner, Elmasonic Xtra ST 300 H/500 H
Ultrasonic Cleaner, Elmasonic Xtra ST 600 H/800 H
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
08-000-121 Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H
08-000-118 Ultrasonic Cleaner, Elmasonic Xtra ST 300 H/500 H
08-000-120 Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H/1600 H
08-000-119 Ultrasonic Cleaner, Elmasonic Xtra ST 600 H/800 H
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 08-000-121 Supplier Elma™ Supplier No. 107 6202
Add to Cart
Add to Cart
For use with Elmasonic Xtra ST Ultrasonic Cleaner.
  • Stainless steel construction
  • Helps to reduce noise and evaporation, serves as drip trays
  • Insulated for sound reduction
Specifications
Product Type Lid
For Use With (Equipment) Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H
Style Hinged Lid
For Use With (Application) Helps to Reduce Noise and Evaporation, Serves as Drip Trays
Material Stainless Steel