Learn More
Electron Microscopy Sciences HINGED PLASTIC VIALS 71.30ML
SDP
Catalog No. 502413124
Encompass_Preferred
These one piece hinged plastic round vials are made from Medical-Grade Polypropylene. Perfect in labs settings for dry or wet samples. They are air and moisture resistant. Have non-allergenis animal-free components and autoclavable (sterilizable) in the presence of aqueous solutions. Food packaging safe. Available in multiple sizes.
Catalog No. 50-241-3124 Supplier Electron Microscopy Sciences Supplier No. 7261722D
These one piece hinged plastic round vials are made from Medical-Grade Polypropylene. Perfect in labs settings for dry or wet samples. They are air and moisture resistant. Have non-allergenis animal-free components and autoclavable (sterilizable) in the presence of aqueous solutions. Food packaging safe. Available in multiple sizes.