Learn More
Electron Microscopy Sciences HINGED RECT PLSTIC BX 33.41ML
SDP
Catalog No. 502413135
Encompass_Preferred
These one piece air and moisture resistant lids are perfect in lab settings for dry or wet samples. With non-allergenis animal-free components are also autoclavable (sterilizable) in the presence of aqueous solutions. Availbale in a variety of sizes, food packaging safe, and recyclable.
Catalog No. 50-241-3135 Supplier Electron Microscopy Sciences Supplier No. 7261761
These one piece air and moisture resistant lids are perfect in lab settings for dry or wet samples. With non-allergenis animal-free components are also autoclavable (sterilizable) in the presence of aqueous solutions. Availbale in a variety of sizes, food packaging safe, and recyclable.