Learn More
Electron Microscopy Sciences HINGED RECT PLSTIC BX 7.39ML
SDP
Supplier:  Electron Microscopy Sciences 7261761M
Encompass_Preferred
These one piece air and moisture resistant lids are perfect in lab settings for dry or wet samples. With non-allergenis animal-free components are also autoclavable (sterilizable) in the presence of aqueous solutions. Availbale in a variety of sizes, food packaging safe, and recyclable.
Catalog No. 50-241-3122