Learn More
Cayman Chemical Hinokiflavone, Purity: ≥95%, CAS: 19202-36-9, 10 mg
SDP
Catalog No. 502911681
Encompass_Preferred
A biflavonoid with diverse biological activities; inhibits the aggregation of Aβ40 in vitro (IC50 = 9.5 μM); promotes pre-mRNA splicing of the genes encoding Mcl-1 and eIF4α and inhibits pre-mRNA splicing of the genes encoding Hsp40 and RIOK3 in NB4 cells at 10 µM; increases protein SUMOylation in HEK293 cells at 10 µM; reduces the cytopathic effect of the influenza A subtypes H1N1 and H1N3, influenza B, parainfluenza type 3, and RSV (IC50s = 1.4, 1.3, 1.2, 1, and 2.5 µg/ml, respectively); inhibits the migration and invasion of, and induces apoptosis in, KYSE-150 cells from 10-40 µM; reduces tumor volume in a KYSE-150 mouse xenograft model at 25 or 50 mg/kg
Catalog No. 50-291-1681 Supplier Cayman Chemical Supplier No. 3633810 MG
A biflavonoid with diverse biological activities; inhibits the aggregation of Aβ40 in vitro (IC50 = 9.5 μM); promotes pre-mRNA splicing of the genes encoding Mcl-1 and eIF4α and inhibits pre-mRNA splicing of the genes encoding Hsp40 and RIOK3 in NB4 cells at 10 µM; increases protein SUMOylation in HEK293 cells at 10 µM; reduces the cytopathic effect of the influenza A subtypes H1N1 and H1N3, influenza B, parainfluenza type 3, and RSV (IC50s = 1.4, 1.3, 1.2, 1, and 2.5 µg/ml, respectively); inhibits the migration and invasion of, and induces apoptosis in, KYSE-150 cells from 10-40 µM; reduces tumor volume in a KYSE-150 mouse xenograft model at 25 or 50 mg/kg