Learn More
Medchemexpress LLC HY-N2360 1mg Medchemexpress, Hinokiflavone CAS:19202-36-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN23601MG
Encompass_Preferred
Medchemexpress, HY-N2360 1mg Hinokiflavone CAS:19202-36-9 Hinokiflavone is a novel modulator of pre-mRNA splicing activity in vitro and in cellulo. Hinokiflavone blocks splicing of pre-mRNA substrates by inhibiting spliceosome assembly, specifically preventing B complex formation. Hinokiflavone is a SUMO protease inhibitor, inhibiting sentrin-specific protease 1 (SENP1) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1498