Learn More
Medchemexpress LLC HY-B2230 100mg Medchemexpress, Hinokitiol CAS:499-44-5 Purity:>98%
SDP
Catalog No. 501933119
Encompass_Preferred
Medchemexpress, HY-B2230 100mg Hinokitiol CAS:499-44-5 Hinokitiol is a component of essential oils isolated from Chymacyparis obtusa, reduces Nrf2 expression, and decreases DNMT1 and UHRF1 mRNA and protein expression, with anti-infective, anti-oxidative, and anti-tumor activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3119 Supplier Medchemexpress LLC Supplier No. HYB2230100MG
Medchemexpress, HY-B2230 100mg Hinokitiol CAS:499-44-5 Hinokitiol is a component of essential oils isolated from Chymacyparis obtusa, reduces Nrf2 expression, and decreases DNMT1 and UHRF1 mRNA and protein expression, with anti-infective, anti-oxidative, and anti-tumor activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.