Learn More
Abcam Hinokitiol, Iron chelator antio x idant, 250MG
Catalog No. 502705574
Encompass_Preferred
MW 164.2 Da, Purity >98%. Iron chelator antioxidant. Inhibits UVB-induced apoptosis. Cytotoxic to cancer cells. Antioxidant, anti-inflammatory and antitumor. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5574 Supplier Abcam Supplier No. AB144606250MG
MW 164.2 Da, Purity >98%. Iron chelator antioxidant. Inhibits UVB-induced apoptosis. Cytotoxic to cancer cells. Antioxidant, anti-inflammatory and antitumor. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement