Learn More
Abcam Hinokitiol, Iron chelator antio x idant, 50MG
Catalog No. 502705573
Encompass_Preferred
MW 164.2 Da, Purity >98%. Iron chelator antioxidant. Inhibits UVB-induced apoptosis. Cytotoxic to cancer cells. Antioxidant, anti-inflammatory and antitumor. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5573 Supplier Abcam Supplier No. AB14460650MG
MW 164.2 Da, Purity >98%. Iron chelator antioxidant. Inhibits UVB-induced apoptosis. Cytotoxic to cancer cells. Antioxidant, anti-inflammatory and antitumor. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement