Learn More
New England Biolabs, Inc. HinP1I – 2000 units
SDP
Catalog No. 50811969
Encompass
A restriction endonuclease that recognizes the sequence G^CG_C.
Catalog No. 50-811-969 Supplier New England Biolabs, Inc. Supplier No. R0124S
A restriction endonuclease that recognizes the sequence G^CG_C.