Learn More
Aviva Systems Biology Corp HINT1 Antibody - N-terminal region (ARP54767_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54767_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HINT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HINT1. Alternate names: FLJ30414, FLJ32340, HINT, PKCI-1, PRKCNH1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61306