Learn More
CritiCore Protective Wear Hip-Length Fluid Barrier Lab Coat
Catalog No. 17012875
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-012-875 5X-Large
17-012-867 X-Small
17-012-868 Small
17-012-869 Medium
17-012-870 Large
17-012-871 X-Large
17-012-872 2X-Large
17-012-873 3X-Large
17-012-874 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 17-012-875 Supplier CritiCore Protective Wear Supplier No. 12L26291BARE015XLWHT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hip-length fluid barrier lab coat constructed with a fluid-repellant front and a breathable, 100% polyester back to optimize protection and comfort.
  • Knit Cuff Elastic Sleeves
  • Concealed Snap Front
  • 3 Pockets
  • Barrier Front
  • 100% Polyester Back
  • Hip Length
  • Primary Applications: Medical/Lab, Dental, Food Processing, Veterinary, Janitorial / Sanitation, Environmental Response
Specifications
Size 5X-Large
Gender Unisex
Material Polyester
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Particulates
Style Regular
No. of Pockets 3