Learn More
Bachem Hippuryl-Lys-OH
SDP
Supplier:  Bachem 4001040.0005
Encompass_Preferred
Hippuryl-Lys-OH, 740-63-6, C15H21N3O4, Mr 307.35, 5g. Bz-Gly-Lys-OH. Hip-Lys, substrate for carboxypeptidase B and carboxypeptidase N.
Catalog No. 50-194-7014