Learn More
MSC Momentive Performance Materials RTV108 3TG 2.8 oz Tube Clear RTV Silicone Joint Sealant 400°F Max Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series RTV100
Catalog No. 501113822
Encompass
Momentive Performance Materials RTV108 3TG 2.8 oz Tube Clear RTV Silicone Joint Sealant 400°F Max Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series RTV100
Catalog No. 50-111-3822 Supplier MSC Supplier No. 06904635
Momentive Performance Materials RTV108 3TG 2.8 oz Tube Clear RTV Silicone Joint Sealant 400°F Max Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series RTV100