Learn More
MSC Momentive Performance Materials RTV102 12C 10.1 oz Tube White RTV Silicone Joint Sealant 204.44°F Max Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series RTV100
Catalog No. 501305167
Encompass
Momentive Performance Materials RTV102 12C 10.1 oz Tube White RTV Silicone Joint Sealant 204.44°F Max Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series RTV100
Catalog No. 50-130-5167 Supplier MSC Supplier No. 06904601
Momentive Performance Materials RTV102 12C 10.1 oz Tube White RTV Silicone Joint Sealant 204.44°F Max Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series RTV100