Learn More
Cole-Parmer Oakton "All-in-one" pH Electrode, single-junction, epoxy-body, gel-filled
Catalog No. 501961784 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. 50-196-1784 Supplier Cole-Parmer Supplier No. 3580176
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.