Learn More
Aviva Systems Biology Corp HIPK1 Antibody - middle region (ARP55866_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55866_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIPK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HIPK1. Alternate names: KIAA0630, MGC26642, MGC33446, MGC33548, Myak, Nbak2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61639