Learn More
Eurofins Discoverx HIPK2, active, 10 µg. (14-623)
Supplier:  Eurofins Discoverx 14623
Encompass
N-terminal 6xHis-tagged recombinant, human HIPK2 amino acids 165-564. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9073