Learn More
Aviva Systems Biology Corp HIPK2 Antibody - middle region (ARP32586_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32586_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HIPK2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human HIPK2. Alternate names: DKFZp686K02111, FLJ23711, PRO0593ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54664