Learn More
Carnabio Usa Inc HIPK3/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 041375
Encompass
HIPK3, Catalytic domain, Wild type, Amino Acid 161-562, NP_005725.3, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0460