Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HIPK3 (YAK1), His Tag Recombinant Protein

Catalog No. PV4324
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4324 1 mg
PR7715A 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV4324 Supplier Invitrogen™ Supplier No. PV4324
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic

HIPK3 is a serine/threonine kinase involved in signal transduction, cellular communication, and multidrug resistance of tumors and plays a role in the phosphorylation of FADD. HIPK3 negatively regulates apoptosis by promoting FADD phosphorylation. This kinase enhances androgen receptor-mediated transcription, and may act as a transcriptional corepressor for NK homeodomain transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H422
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 150mM NaCl, 0.02% Triton X-100, 50% glycerol, 2mM DTT, 0.5mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 10114
Molecular Weight (g/mol) 49.7 kDa
Name Human HIPK3 (YAK1), His Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Androgen receptor-interacting nuclear protein kinase; ANPK; DYRK6; Fas-interacting serine/threonine-protein kinase; FIST; FIST3; HCMV-interacting protein; HIPK3; homeodomain interacting protein kinase 3; homeodomain-interacting protein kinase 3; Homolog of protein kinase YAK1; mir-1902; nbak3; Nuclear body-associated kinase 3; nuclear serine/threonine protein kinase; PKY; RP1-8L15.1; YAK1
Common Name HIPK3
Gene Symbol HIPK3
Kinase Family DYRK Kinase Family
Protein Length 163-562
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence 163-562
Content And Storage Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Kinase Group Ser⁄Thr Kinases: CMGC Group
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Catalytic
Form Liquid
Research Category Drug Development
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.