Learn More
Cell Signaling Technology Hippo Signaling Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502730664
Encompass
The Hippo Signaling Antibody Sampler Kit provides an economical means of detecting target proteins of the Hippo signaling pathway. The kit contains enough primary antibody to perform two western blots per primary.
  • Contains an assortment of antibodies targeting key proteins in the Hippo signaling pathway, including YAP, TAZ, MOB1, LATS1, MST1, MST2, and SAV1
  • Supplied with Anti-rabbit IgG, HRP-linked secondary antibody
  • Ideal for analyzing the activation status of the Hippo pathway
  • Includes primary antibodies against phosphorylated and total protein targets
Catalog No. 50-273-0664 Supplier Cell Signaling Technology Supplier No. 8579T
The Hippo Signaling Antibody Sampler Kit provides an economical means of detecting target proteins of the Hippo signaling pathway. The kit contains enough primary antibody to perform two western blots per primary.
  • Contains an assortment of antibodies targeting key proteins in the Hippo signaling pathway, including YAP, TAZ, MOB1, LATS1, MST1, MST2, and SAV1
  • Supplied with Anti-rabbit IgG, HRP-linked secondary antibody
  • Ideal for analyzing the activation status of the Hippo pathway
  • Includes primary antibodies against phosphorylated and total protein targets