Learn More
Selleck Chemical LLC Hippuric acid
SDP
Catalog No. 5000003208
Encompass_Preferred
Hippuric Acid (2-Benzamidoacetic acid Benzoylglycine) is an acyl glycine produced by the conjugation of benzoic acid and glycine found as a normal component in urine as a metabolite of aromatic compounds from food
Catalog No. 50-000-03208 Supplier Selleck Chemical LLC Supplier No. S561825MG
Hippuric Acid (2-Benzamidoacetic acid Benzoylglycine) is an acyl glycine produced by the conjugation of benzoic acid and glycine found as a normal component in urine as a metabolite of aromatic compounds from food