Learn More
Medchemexpress LLC Hippuric acid | 495-69-2 | MFCD00002692 | 100.0% | 179.18 | C9H9NO3 | 5 G
SDP
Catalog No. 5000299365
Encompass_Preferred
Hippuric acid (CAS 495-69-2; C9H9NO3; MW 179.18) is an endogenous metabolite supplied as a research-grade reagent for biochemical and metabolic studies. Provided as a stable solid, it can be formulated for in vitro and in vivo experiments following manufacturer solubility guidance. For research use only.
  • High purity (99.98%).
  • Soluble in DMSO: 75 mg/mL (requires ultrasonic).
  • Able to form clear in vivo formulations at ≥3.75 mg/mL in recommended vehicles.
  • Stable storage: room temperature (solid) up to 3 years; in solution, store at -80°C or -20°C per guidance.
  • Suitable as a metabolite reference and for biochemical or pathway studies.
Catalog No. 50-002-99365 Supplier Medchemexpress LLC Supplier No. HYW0165625G
Hippuric acid (CAS 495-69-2; C9H9NO3; MW 179.18) is an endogenous metabolite supplied as a research-grade reagent for biochemical and metabolic studies. Provided as a stable solid, it can be formulated for in vitro and in vivo experiments following manufacturer solubility guidance. For research use only.
  • High purity (99.98%).
  • Soluble in DMSO: 75 mg/mL (requires ultrasonic).
  • Able to form clear in vivo formulations at ≥3.75 mg/mL in recommended vehicles.
  • Stable storage: room temperature (solid) up to 3 years; in solution, store at -80°C or -20°C per guidance.
  • Suitable as a metabolite reference and for biochemical or pathway studies.