Learn More
Medchemexpress LLC Hippuric acid | 495-69-2 | MFCD00002692 | 100.0% | 179.18 | C9H9NO3 | 500 MG
SDP
Catalog No. 5000299485
Encompass_Preferred
Hippuric acid is an endogenous metabolite (benzoylglycine) commonly used as a biochemical research reagent and analytical standard. It is applied in studies of metabolism, toxicology, and signaling, and its chemical properties and storage recommendations are documented for laboratory use.
  • Endogenous metabolite used in metabolic and biomarker research.
  • Relevant to studies of oxidative stress, MMP/NRF2 pathways, and TGFβ/SMAD signaling.
  • Suitable as an analytical reference standard and biochemical reagent.
  • Stable under recommended storage: room temperature for the solid; long-term storage in solvent at -80°C.
  • Well-characterized chemical properties: CAS 495-69-2, formula C9H9NO3, mw 179.18.
Catalog No. 50-002-99485 Supplier Medchemexpress LLC Supplier No. HYW016562500MG
Hippuric acid is an endogenous metabolite (benzoylglycine) commonly used as a biochemical research reagent and analytical standard. It is applied in studies of metabolism, toxicology, and signaling, and its chemical properties and storage recommendations are documented for laboratory use.
  • Endogenous metabolite used in metabolic and biomarker research.
  • Relevant to studies of oxidative stress, MMP/NRF2 pathways, and TGFβ/SMAD signaling.
  • Suitable as an analytical reference standard and biochemical reagent.
  • Stable under recommended storage: room temperature for the solid; long-term storage in solvent at -80°C.
  • Well-characterized chemical properties: CAS 495-69-2, formula C9H9NO3, mw 179.18.