Learn More
Medchemexpress LLC Hippuric acid-d2 | 208928-78-3 | MFCD01317354 | 99.8% | 181.18 g·mol⁻1 | C9H7D2NO3 | 10 MG
SDP
Catalog No. 5000301147
Encompass_Preferred
Hippuric acid-d2 is a deuterium-labeled analogue of hippuric acid used as a stable isotope internal standard for quantitative mass spectrometry and metabolite identification. It provides a predictable mass shift for isotope-dilution assays, supporting accurate quantitation and method development.
  • Deuterium-labeled internal standard for LC-MS/MS quantitation.
  • Enables isotope-dilution assays with improved accuracy and precision.
  • High purity (99.8%) for reliable analytical results.
  • Available in small amounts suitable for method development and calibration.
  • Solid form, stable under normal laboratory storage conditions.
Catalog No. 50-003-01147 Supplier Medchemexpress LLC Supplier No. HYW016562S110MG
Hippuric acid-d2 is a deuterium-labeled analogue of hippuric acid used as a stable isotope internal standard for quantitative mass spectrometry and metabolite identification. It provides a predictable mass shift for isotope-dilution assays, supporting accurate quantitation and method development.
  • Deuterium-labeled internal standard for LC-MS/MS quantitation.
  • Enables isotope-dilution assays with improved accuracy and precision.
  • High purity (99.8%) for reliable analytical results.
  • Available in small amounts suitable for method development and calibration.
  • Solid form, stable under normal laboratory storage conditions.