Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hippuric acid-d2 | 208928-78-3 | MFCD01317354 | 99.8% | 181.18 g/mol | C9H7D2NO3 | 25 MG
SDP

Catalog No. 5000303407
Encompass_Preferred

Hippuric acid-d2 is a deuterium-labeled reference standard (2-benzamidoacetic acid-d2) supplied as a high-purity solid for use as an internal standard or tracer in LC-MS/MS, metabolic profiling, and related analytical workflows. It provides defined isotopic labeling to improve accuracy and precision in quantitation.

  • Deuterium-labeled internal standard for mass spectrometry assays.
  • High purity (99.83%) ensures consistent quantitation.
  • Molecular weight 181.18 g/mol and formula C9H7D2NO3 for unambiguous identification.
  • Solid, white to off-white form for stable storage and handling.
  • Recommended storage: -20°C (powder) or -80°C (in solvent) to minimize degradation.

Catalog No. 50-003-03407 Supplier Medchemexpress LLC Supplier No. 25MGHYW016562S1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hippuric acid-d2 is a deuterium-labeled reference standard (2-benzamidoacetic acid-d2) supplied as a high-purity solid for use as an internal standard or tracer in LC-MS/MS, metabolic profiling, and related analytical workflows. It provides defined isotopic labeling to improve accuracy and precision in quantitation.

  • Deuterium-labeled internal standard for mass spectrometry assays.
  • High purity (99.83%) ensures consistent quantitation.
  • Molecular weight 181.18 g/mol and formula C9H7D2NO3 for unambiguous identification.
  • Solid, white to off-white form for stable storage and handling.
  • Recommended storage: -20°C (powder) or -80°C (in solvent) to minimize degradation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.