Learn More
Medchemexpress LLC Hippuric acid-d2 | 208928-78-3 | MFCD01317354 | 99.8% | 181.18 g/mol | C9H7D2NO3 | 5 MG
SDP
Catalog No. 5000299364
Encompass_Preferred
Hippuric acid-d2 is the deuterium-labeled form of hippuric acid intended for use as a tracer and internal standard in quantitative analytical methods. It is supplied in small milligram quantities and characterized for isotopic and chemical purity to support accurate NMR, GC-MS, and LC-MS measurements.
  • Deuterium-labeled analog for isotope dilution assays.
  • High purity for reliable quantitative results.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Provided in small milligram quantities for analytical workflows.
  • Documentation available including COA, SDS, and LCMS report.
Catalog No. 50-002-99364 Supplier Medchemexpress LLC Supplier No. HYW016562S15MG
Hippuric acid-d2 is the deuterium-labeled form of hippuric acid intended for use as a tracer and internal standard in quantitative analytical methods. It is supplied in small milligram quantities and characterized for isotopic and chemical purity to support accurate NMR, GC-MS, and LC-MS measurements.
  • Deuterium-labeled analog for isotope dilution assays.
  • High purity for reliable quantitative results.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Provided in small milligram quantities for analytical workflows.
  • Documentation available including COA, SDS, and LCMS report.