Learn More
Medchemexpress LLC Hippuric acid (Standard) | 495-69-2 | MFCD00002692 | 99.8% | 179.17 g/mol | C9H9NO3 | 50 MG
SDP
Catalog No. 5000299366
Encompass_Preferred
Hippuric acid (Standard) is an analytical/reference standard of benzoylglycine intended for use in analytical chemistry, method development, and metabolite studies. Supplied in small laboratory pack sizes, it is intended for calibration, quantitative analysis, and chromatographic applications.
  • Analytical reference standard for calibration and method development.
  • High purity for analytical use (≈99.8%).
  • Molecular formula C9H9NO3; molecular weight 179.17 g/mol.
  • Supplied in small milligram pack sizes for laboratory use.
  • Suitable for chromatography and metabolite quantification.
Catalog No. 50-002-99366 Supplier Medchemexpress LLC Supplier No. HYW016562R50MG
Hippuric acid (Standard) is an analytical/reference standard of benzoylglycine intended for use in analytical chemistry, method development, and metabolite studies. Supplied in small laboratory pack sizes, it is intended for calibration, quantitative analysis, and chromatographic applications.
  • Analytical reference standard for calibration and method development.
  • High purity for analytical use (≈99.8%).
  • Molecular formula C9H9NO3; molecular weight 179.17 g/mol.
  • Supplied in small milligram pack sizes for laboratory use.
  • Suitable for chromatography and metabolite quantification.