Learn More
Medchemexpress LLC Hippuric acid (standard) | 495-69-2 | MFCD00002692 | 99.8% | 179.18 | C9H9NO3 | 25 MG
SDP
Catalog No. 5000303043
Encompass_Preferred
Hippuric acid (standard) is an analytical reference material of hippuric acid supplied as a solid with verified identity and documentation. It is intended for laboratory research use such as metabolite quantification, method validation, and quality control, and includes storage guidance and traceable purity data.
  • Verified purity (99.8%) suitable for quantitative analysis.
  • Solid form for easy handling and accurate weighing.
  • Stable under recommended storage conditions for reliable results.
  • Suitable for metabolite quantification, method development, and quality control.
  • Includes supporting documentation (SDS, datasheet, COA) for traceability.
Catalog No. 50-003-03043 Supplier Medchemexpress LLC Supplier No. 25MGHYW016562R
Hippuric acid (standard) is an analytical reference material of hippuric acid supplied as a solid with verified identity and documentation. It is intended for laboratory research use such as metabolite quantification, method validation, and quality control, and includes storage guidance and traceable purity data.
  • Verified purity (99.8%) suitable for quantitative analysis.
  • Solid form for easy handling and accurate weighing.
  • Stable under recommended storage conditions for reliable results.
  • Suitable for metabolite quantification, method development, and quality control.
  • Includes supporting documentation (SDS, datasheet, COA) for traceability.