Learn More
Sigma Aldrich Fine Chemicals Biosciences HiPrepTM 26 0 Desalting C
Catalog No. 502908074
Encompass
HiPrep(TM) 26/10 Desalting is a convenient easy-to-use column prepacked 53 mL column with Sephadex(R) G-25 Fine for desalting and buffer exchange.
Catalog No. 50-290-8074 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17508701
Add to Cart
Add to Cart
HiPrep(TM) 26/10 Desalting is a convenient easy-to-use column prepacked 53 mL column with Sephadex(R) G-25 Fine for desalting and buffer exchange.