Learn More
Sigma Aldrich Fine Chemicals Biosciences HiPrepTM 26/60 SephacrylR
Catalog No. 502908198
Encompass
HiPrep(TM) 26/60 Sephacryl(R) S-100 HR are prepacked gel filtration columns for preparative separation of biomolecules from up to 13 mL sample volume. Sephacryl(R) S-100 is used for separating peptides and small proteins.
Catalog No. 50-290-8198 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17119401
Add to Cart
Add to Cart
HiPrep(TM) 26/60 Sephacryl(R) S-100 HR are prepacked gel filtration columns for preparative separation of biomolecules from up to 13 mL sample volume. Sephacryl(R) S-100 is used for separating peptides and small proteins.