Learn More
Sigma Aldrich Fine Chemicals Biosciences HiPrepTM Heparin Fast Flow
Catalog No. 502907432
Encompass
HiPrep(TM) Heparin FF 16/10 is prepacked with Heparin Sepharose(R) 6 Fast FLow for preparative purification of proteins with affinity for heparin such as DNA-binding proteins coagulation factors lipoproteins and protein synthesis factors.
Catalog No. 50-290-7432 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28936549
HiPrep(TM) Heparin FF 16/10 is prepacked with Heparin Sepharose(R) 6 Fast FLow for preparative purification of proteins with affinity for heparin such as DNA-binding proteins coagulation factors lipoproteins and protein synthesis factors.