Learn More
Sigma Aldrich Fine Chemicals Biosciences HiPrepTM Q Fast Flow 16 0
Catalog No. 502907120
Encompass
HiPrep(TM) Q FF 16/10 is a strong anion exchanger prepacked with Q Sepharose(R) Fast Flow ready-to-use for fast preparative separations of proteins and other biomolecules using ion exchange (IEX) chromatography.HiPrep(TM) Q FF are prepacked ready to use columns for ion exchange chromatography. They provide fast preparative separations of proteins and other biomolecules.
Catalog No. 50-290-7120 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28936543
HiPrep(TM) Q FF 16/10 is a strong anion exchanger prepacked with Q Sepharose(R) Fast Flow ready-to-use for fast preparative separations of proteins and other biomolecules using ion exchange (IEX) chromatography.HiPrep(TM) Q FF are prepacked ready to use columns for ion exchange chromatography. They provide fast preparative separations of proteins and other biomolecules.