Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ HiPur™ Denatured Ethyl Alcohol

Catalog No. 22110882 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-110-882 5 gal.
22-110-880 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-110-882 Supplier Epredia™ Supplier No. 92005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Choose the option that best fits your work, HiPur™ Denatured Ethanol is available in one or five gallon sizes.

  • Maximum of 0.1% water
  • Maximum residue content of 0.003%
  • Anhydrous formula is denatured with Methanol, MIBK, Ethyl Acetate and Heptane

Specifications

Product Type Alcohol
Quantity 5 gal.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.