Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HiPur™ Formaldehyde (37%), Epredia™

Catalog No. 22110870 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
Packaging:
HDPE Bottle
Poly Carboy
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
22-110-870 1 gal. HDPE Bottle
22-110-872 5 gal. Poly Carboy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-110-870 Supplier Epredia Supplier No. 93001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Solution is stabilized with Methanol. Not intended for use as a disinfectant.

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
IUPAC Name formaldehyde
SMILES C=O

Specifications

Packaging HDPE Bottle
Quantity 1 gal.
Concentration or Composition (by Analyte or Components) 0.37
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.