Learn More
Aviva Systems Biology Corp Hira Antibody - middle region (ARP37054_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37054_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hira. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse Hira. Alternate names: AA138857, D16Ertd95e, N28177, Tuple1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-574