Learn More
Chiral Technologies, Inc. Chiralpak IE-3 2.1Mm 3Um Column, High Selectivity, Solvent Versatility, Ready-To-Use
Catalog No. NC2743059
Encompass
The CHIRALPAK IE-3 2.1 x 100 mm 3 µm column is designed for high-performance chiral separation in HPLC and SFC. Featuring an immobilized polysaccharide stationary phase, it ensures exceptional solvent compatibility and enhanced selectivity, making it suitable for demanding analytical methods. Its smaller particle size enables fast resolution and easy method transfer from larger columns, differentiating it in the market.
Catalog No. NC2743059 Supplier Chiral Technologies, Inc. Supplier No. 85593
The CHIRALPAK IE-3 2.1 x 100 mm 3 µm column is designed for high-performance chiral separation in HPLC and SFC. Featuring an immobilized polysaccharide stationary phase, it ensures exceptional solvent compatibility and enhanced selectivity, making it suitable for demanding analytical methods. Its smaller particle size enables fast resolution and easy method transfer from larger columns, differentiating it in the market.