Learn More
Roche Diagnostics HIRES REAGENT KIT
Supplier:  Roche Diagnostics 05650798001
Encompass
The Cedex HiRes Reagent Kit contains all reagents required for preparation, staining, and analysis of eukaryotic cells using the Cedex HiRes System. In addition, a waste container is provided for liquid waste.
Catalog No. 50-589-363