Learn More
Andwin Scientific HIRSCHMANN PIPETTUS
SDP
Catalog No. NC0175827
Encompass_Preferred
The pipetus pipet filler features a color display that shows the battery level, set pump speed, and dispensing mode. It also has two reusable, built-in safety check valve that acts as a first line of defense from media being accidentally drawn up too far. This product is the only pipet filler with an inductive charging stand that can also perform as a pipet holder.
Catalog No. NC0175827 Supplier Andwin Scientific Supplier No. 83007692
Add to Cart
Add to Cart
The pipetus pipet filler features a color display that shows the battery level, set pump speed, and dispensing mode. It also has two reusable, built-in safety check valve that acts as a first line of defense from media being accidentally drawn up too far. This product is the only pipet filler with an inductive charging stand that can also perform as a pipet holder.